Gene Transcripts

Gene Symbol
FBXL22
Entrez Gene ID
283807
Gene Name
F-box and leucine rich repeat protein 22
Gene Aliases
Fbl22
Chromosome Location
Chr. 15: 63596225 - 63608898 on Build GRCh38
UniGene ID
Hs.631163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203373.2 NP_976307.2 2 1376
GenBank mRNA BC065833.1 - 2 1332

View Full Product Details