Gene Transcripts

Gene Symbol
IZUMO1
Entrez Gene ID
284359
Gene Name
izumo sperm-egg fusion 1
Gene Aliases
IZUMO, OBF
Chromosome Location
Chr. 19: 48740888 - 48747499 on Build GRCh38
UniGene ID
Hs.400688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321864.1 NP_001308793.1 8, 9 873
NM_001321865.1 NP_001308794.1 8, 9 1036
NM_182575.2 NP_872381.2 9, 10 1474
NR_135832.1 8, 9 962
XM_005258793.4 XP_005258850.1 9, 10 1207
XM_011526811.2 XP_011525113.1 7, 8 1897
XM_011526812.2 XP_011525114.1 9, 10 1554
XM_011526813.2 XP_011525115.1 9, 10 1403
XM_011526815.2 XP_011525117.1 8, 9 2117
XM_017026646.1 XP_016882135.1 8, 9 1978
GenBank mRNA AB195682.1 - 9, 10 1332
AK301750.1 - 8, 9 2117
AY552609.1 - 8, 9 963
AY552610.1 - 8, 9 889
BC034769.1 - 9, 10 1474 5167853

View Full Product Details