Gene Transcripts

Gene Symbol
IZUMO1
Entrez Gene ID
284359
Gene Name
izumo sperm-egg fusion 1
Gene Aliases
IZUMO, OBF
Chromosome Location
Chr. 19: 48740888 - 48747499 on Build GRCh38
UniGene ID
Hs.400688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321864.1 NP_001308793.1 9 906
NM_001321865.1 NP_001308794.1 9 1069
NM_182575.2 NP_872381.2 10 1507
NR_135832.1 9 995
XM_005258793.4 XP_005258850.1 10 1240
XM_011526811.2 XP_011525113.1 8 1930
XM_011526812.2 XP_011525114.1 10 1587
XM_011526813.2 XP_011525115.1 10 1436
XM_011526815.2 XP_011525117.1 9 2150
XM_017026646.1 XP_016882135.1 9 2011
GenBank mRNA AB195682.1 - 10 1365
AK301750.1 - 9 2150
AY552609.1 - 9 996
AY552610.1 - 9 922
BC034769.1 - 10 1507 5167853

View Full Product Details