Gene Transcripts

Gene Symbol
DPH3
Entrez Gene ID
285381
Gene Name
diphthamide biosynthesis 3
Gene Aliases
DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2
Chromosome Location
Chr. 3: 16257061 - 16264989 on Build GRCh38
UniGene ID
Hs.388087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047434.2 NP_001040899.1 2 453
NM_206831.2 NP_996662.1 3 528
GenBank mRNA AK022970.1 - 2 433
AK092040.1 - 3 481
AK311618.1 - 3 490
BC010181.2 - 3 437 4543414
DA447430.1 - 2 443
DA875039.1 - 3 528

View Full Product Details