Gene Transcripts

Gene Symbol
FAM78A
Entrez Gene ID
286336
Gene Name
family with sequence similarity 78 member A
Gene Aliases
C9orf59
Chromosome Location
Chr. 9: 131258078 - 131281930 on Build GRCh38
UniGene ID
Hs.143878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033387.3 NP_203745.2 1 458
GenBank mRNA AK095423.1 - 1 486
BC049388.2 - 1 458 6050655

View Full Product Details