Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000257.3 | NP_000248.2 | 10 | 951 | |
XM_017021340.1 | XP_016876829.1 | 9 | 886 | ||
GenBank mRNA | AK307590.1 | - | 10 | 925 | |
BC112171.1 | - | 9 | 875 | 8327639 | |
BC112173.1 | - | 9 | 875 | 8327641 | |
EF560725.1 | - | 8 | 818 | ||
EU747717.1 | - | 10 | 926 | ||
M58018.1 | - | 10 | 906 |