Gene Transcripts

Gene Symbol
KRTAP7-1
Entrez Gene ID
337878
Gene Name
keratin associated protein 7-1 (gene/pseudogene)
Gene Aliases
KAP7.1
Chromosome Location
Chr. 21: 30829039 - 30829732 on Build GRCh38
UniGene ID
Hs.407656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181606.2 NP_853637.2 1 488
GenBank mRNA AJ457063.1 - 1 365

View Full Product Details