Gene Transcripts

Gene Symbol
OR5AP2
Entrez Gene ID
338675
Gene Name
olfactory receptor family 5 subfamily AP member 2
Gene Aliases
OR9J1
Chromosome Location
Chr. 11: 56641489 - 56642439 on Build GRCh38
UniGene ID
Hs.449692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002925.1 NP_001002925.1 1 809
GenBank mRNA BC136989.1 - 1 841
BC136990.1 - 1 841 9020982

View Full Product Details