Gene Transcripts

Gene Symbol
C17orf51
Entrez Gene ID
339263
Gene Name
chromosome 17 open reading frame 51
Gene Aliases
Chromosome Location
Chr. 17: 21524606 - 21551678 on Build GRCh38
UniGene ID
Hs.514016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113434.4 NP_001106905.1 1 644
XM_005256621.3 XP_005256678.1 1 637
XM_006721514.2 XP_006721577.1 1 637
XM_011523822.2 XP_011522124.1 1 637
GenBank mRNA AK056436.1 - 1 610
BC010612.1 - 1 642
BC019938.1 - 1 632
BC052777.1 - 1 644
BC053952.1 - 1 644
BC132738.1 - 1 618 40146760
BC136630.1 - 1 618 9020620

View Full Product Details