Gene Transcripts

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
Chromosome Location
Chr. 19: 7688757 - 7702755 on Build GRCh38
UniGene ID
Hs.465778
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207019.2 NP_001193948.2 8 600
NM_001220500.1 NP_001207429.1 9 736
NM_002002.4 NP_001993.2 9 732
XM_005272462.4 XP_005272519.1 9 1337
GenBank mRNA AK223207.1 - 9 701
AK310071.1 - 8 881
BC014108.2 - 9 697 4309266
BC062591.1 - 9 690 5216918
BC064417.1 - 8 1014
M14766.1 - 9 704
M15059.1 - 9 732
X04772.1 - 9 732

View Full Product Details