Gene Transcripts
- Gene Symbol
- OR2W3
- Entrez Gene ID
- 343171
- Gene Name
- olfactory receptor family 2 subfamily W member 3
- Gene Aliases
- OR2W3P, OR2W8P, OST718
- Chromosome Location
- Chr. 1: 247895587 - 247896531 on Build GRCh38
- UniGene ID
- Hs.269151
- Species
- Homo sapiens
- Gene Symbol
- TRIM58
- Entrez Gene ID
- 25893
- Gene Name
- tripartite motif containing 58
- Gene Aliases
- BIA2
- Chromosome Location
- Chr. 1: 247857199 - 247880138 on Build GRCh38
- UniGene ID
- Hs.269151
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537432.1 |
- |
3 |
657 |
|
View Full Product Details