Gene Transcripts

Gene Symbol
ARHGAP40
Entrez Gene ID
343578
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, dJ1100H13.4
Chromosome Location
Chr. 20: 38601934 - 38650652 on Build GRCh38
UniGene ID
Hs.451997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164431.1 NP_001157903.1 9 1184

View Full Product Details