Gene Transcripts

Gene Symbol
MTX3
Entrez Gene ID
345778
Gene Name
metaxin 3
Gene Aliases
Chromosome Location
Chr. 5: 79976716 - 79991267 on Build GRCh38
UniGene ID
Hs.531418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010891.4 NP_001010891.4 5 381
NM_001167741.1 NP_001161213.1 4 311
XM_005248495.3 XP_005248552.1 5 383
XM_017009440.1 XP_016864929.1 4 305
GenBank mRNA AK296864.1 - 5 376
BC150295.1 - 5 371
BX538064.1 - 4 291
BX647596.1 - 5 378
DB456588.1 - 5 381

View Full Product Details