Gene Transcripts

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
Chromosome Location
Chr. 6: 63719980 - 65707225 on Build GRCh38
UniGene ID
Hs.25067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142800.1 NP_001136272.1 11 2190
NM_001142801.1 NP_001136273.1 11 2190
NM_001292009.1 NP_001278938.1 11 2190
NM_198283.1 NP_938024.1 10 2049
GenBank mRNA AB593114.1 - 11 2155
AY358133.1 - 10 2049
BC133011.1 - 9 1914
BC133013.1 - 9 1911
BX537914.1 - 11 2190
FJ416331.1 - 11 2198
FM209056.1 - 11 2142

View Full Product Details