Gene Transcripts

Gene Symbol
LANCL3
Entrez Gene ID
347404
Gene Name
LanC like 3
Gene Aliases
Chromosome Location
Chr. X: 37571569 - 37684463 on Build GRCh38
UniGene ID
Hs.521932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170331.1 NP_001163802.1 2 896
NM_198511.2 NP_940913.1 2 896
XM_011543904.2 XP_011542206.1 2 381
GenBank mRNA AK124915.1 - 2 896
BC093667.1 - 2 688 7939512
BC093669.1 - 2 688 7939514
BC143528.1 - 2 688

View Full Product Details