Gene Transcripts

Gene Symbol
XKRY2
Entrez Gene ID
353515
Gene Name
XK related, Y-linked 2
Gene Aliases
XKRYP7
Chromosome Location
Chr. Y: 18135449 - 18137029 on Build GRCh38
UniGene ID
Hs.632286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002906.1 NP_001002906.1 1 893
GenBank mRNA BC132901.1 - 1 765
BC132903.1 - 1 765 40146925

Gene Symbol
XKRY
Entrez Gene ID
9082
Gene Name
XK related, Y-linked
Gene Aliases
XKRY1
Chromosome Location
Chr. Y: 17768980 - 17770560 on Build GRCh38
UniGene ID
Hs.158346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004677.2 NP_004668.2 1 893
GenBank mRNA AF000997.1 - 1 893
BC132883.1 - 1 765 40146905
BC132885.1 - 1 765 40146907

View Full Product Details