Gene Transcripts
- Gene Symbol
- DNAJB13
- Entrez Gene ID
- 374407
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B13
- Gene Aliases
- RSPH16A, TSARG5, TSARG6
- Chromosome Location
- Chr. 11: 73950319 - 73970287 on Build GRCh38
- UniGene ID
- Hs.567888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153614.3 |
NP_705842.2
|
4, 5 |
1247 |
|
XM_005273984.3 |
XP_005274041.1
|
3, 4 |
612 |
|
XM_011545004.2 |
XP_011543306.1
|
6, 7 |
1498 |
|
XM_011545005.2 |
XP_011543307.1
|
5, 6 |
1338 |
|
XM_011545007.2 |
XP_011543309.1
|
4, 5 |
1023 |
|
XM_011545009.2 |
XP_011543311.1
|
6, 7 |
1172 |
|
XM_011545013.2 |
XP_011543315.1
|
6, 7 |
1504 |
|
XM_017017675.1 |
XP_016873164.1
|
5, 6 |
768 |
|
XM_017017676.1 |
XP_016873165.1
|
6, 7 |
1348 |
|
XM_017017677.1 |
XP_016873166.1
|
5, 6 |
1217 |
|
XM_017017678.1 |
XP_016873167.1
|
4, 5 |
1028 |
|
GenBank mRNA |
AK128589.1 |
- |
6, 7 |
3292 |
|
AY138810.2 |
- |
4, 5 |
1247 |
|
AY325766.1 |
- |
4, 5 |
540 |
|
View Full Product Details