Gene Transcripts

Gene Symbol
KRTAP12-3
Entrez Gene ID
386683
Gene Name
keratin associated protein 12-3
Gene Aliases
KRTAP12.3
Chromosome Location
Chr. 21: 44657932 - 44658341 on Build GRCh38
UniGene ID
Hs.528023
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198697.2 NP_941970.2 1 235
GenBank mRNA AB076361.1 - 1 235

View Full Product Details