Gene Transcripts
- Gene Symbol
- THEMIS
- Entrez Gene ID
- 387357
- Gene Name
- thymocyte selection associated
- Gene Aliases
- C6orf190, C6orf207, GASP, SPOT, TSEPA
- Chromosome Location
- Chr. 6: 127697191 - 127918631 on Build GRCh38
- UniGene ID
- Hs.661756
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001010923.2 |
NP_001010923.1
|
3, 4 |
854 |
|
NM_001164685.1 |
NP_001158157.1
|
3, 4 |
854 |
|
NM_001164687.1 |
NP_001158159.1
|
3, 4 |
818 |
|
NM_001318531.1 |
NP_001305460.1
|
4, 5 |
919 |
|
XM_011535813.2 |
XP_011534115.1
|
5, 6 |
1048 |
|
XM_011535814.2 |
XP_011534116.1
|
4, 5 |
819 |
|
XM_011535816.1 |
XP_011534118.1
|
5, 6 |
1044 |
|
XM_017010848.1 |
XP_016866337.1
|
5, 6 |
1048 |
|
GenBank mRNA |
AK097903.1 |
- |
3, 4 |
831 |
|
AK124031.1 |
- |
3, 4 |
784 |
|
AK128377.1 |
- |
4, 5 |
852 |
|
AK292046.1 |
- |
3, 4 |
804 |
|
BC130516.1 |
- |
3, 4 |
848 |
40146547 |
BC144034.1 |
- |
3, 4 |
846 |
|
View Full Product Details