Gene Transcripts

Gene Symbol
RASL11A
Entrez Gene ID
387496
Gene Name
RAS like family 11 member A
Gene Aliases
Chromosome Location
Chr. 13: 27270327 - 27273690 on Build GRCh38
UniGene ID
Hs.192131
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206827.1 NP_996563.1 2, 3 799
GenBank mRNA AY439004.1 - 2, 3 799
BC132703.1 - 2, 3 535 40146725
BC136761.1 - 2, 3 535 9020751

View Full Product Details