Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207377.2 | NP_997260.1 | 4 | 410 | |
XM_011536742.2 | XP_011535044.1 | 4 | 453 | ||
XM_011536743.2 | XP_011535045.1 | 4 | 456 | ||
XM_011536744.2 | XP_011535046.1 | 2 | 304 | ||
GenBank mRNA | AY358266.1 | - | 4 | 422 | |
BC137557.1 | - | 4 | 408 | 9021557 | |
BC137563.1 | - | 4 | 408 | 9021563 | |
BC144709.1 | - | 3 | 326 |