Gene Transcripts

Gene Symbol
OR11G2
Entrez Gene ID
390439
Gene Name
olfactory receptor family 11 subfamily G member 2
Gene Aliases
OR14-34
Chromosome Location
Chr. 14: 20197336 - 20198373 on Build GRCh38
UniGene ID
Hs.554582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005503.1 NP_001005503.1 1 981

View Full Product Details