Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location
Chr. 14: 88385638 - 88470350 on Build GRCh38
UniGene ID
Hs.525518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040428.3 NP_001035518.1 11 1317
NM_018418.4 NP_060888.2 12 1413
XM_005267851.1 XP_005267908.1 12 1387
XM_005267852.1 XP_005267909.1 11 1316
XM_005267854.1 XP_005267911.1 13 1449
XM_005267855.1 XP_005267912.1 12 1382
XM_006720204.1 XP_006720267.1 12 1387
XM_006720205.1 XP_006720268.1 12 1387
XM_011536951.1 XP_011535253.1 12 1581
XM_011536952.1 XP_011535254.1 10 1199
XM_011536953.1 XP_011535255.1 11 1520
XM_017021452.1 XP_016876941.1 12 1578
XM_017021453.1 XP_016876942.1 12 1547
XM_017021454.1 XP_016876943.1 13 1446
XM_017021455.1 XP_016876944.1 12 1379
XM_017021456.1 XP_016876945.1 13 1392
XM_017021457.1 XP_016876946.1 11 1517
GenBank mRNA AF144487.3 - 12 1393
AF144488.2 - 10 1121
AK055864.1 - 11 2985
AK225746.1 - 13 1387
AK290631.1 - 11 1288
AL136604.1 - 11 1297
BC008656.1 - 7 845
BC034635.1 - 2480
BC090875.1 - 12 1700 30383306
CR533563.1 - 11 1142
CR936777.1 - 12 1359
EU668353.1 - 12 1389

View Full Product Details