Gene Transcripts

Gene Symbol
UNC13D
Entrez Gene ID
201294
Gene Name
unc-13 homolog D
Gene Aliases
FHL3, HLH3, HPLH3, Munc13-4
Chromosome Location
Chr. 17: 75827225 - 75844717 on Build GRCh38
UniGene ID
Hs.41045
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199242.2 NP_954712.1 21 2356
XM_011524504.1 XP_011522806.1 21 1989
XM_011524506.1 XP_011522808.1 21 1986
XM_011524507.2 XP_011522809.1 21 2386
GenBank mRNA AJ578444.1 - 21 2034
AK024474.1 - 19 2258
AK300316.1 - 22 2047
AK301529.1 - 21 1989
BC024740.2 - 19 1785
BC067084.1 - 21 2356 5951944

View Full Product Details