Gene Transcripts

Gene Symbol
SPINT4
Entrez Gene ID
391253
Gene Name
serine peptidase inhibitor, Kunitz type 4
Gene Aliases
C20orf137, SPINT3, dJ601O1.1
Chromosome Location
Chr. 20: 45722349 - 45725696 on Build GRCh38
UniGene ID
Hs.677891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178455.1 NP_848550.1 2 181
GenBank mRNA AY372173.1 - 2 181
AY372174.1 - 2 181

View Full Product Details