Gene Transcripts

Gene Symbol
FAM102A
Entrez Gene ID
399665
Gene Name
family with sequence similarity 102 member A
Gene Aliases
C9orf132, EEIG1, SYM-3A, bA203J24.7
Chromosome Location
Chr. 9: 127940582 - 127980533 on Build GRCh38
UniGene ID
Hs.535972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035254.2 NP_001030331.1 6 917
NM_203305.2 NP_976050.1 3 399
GenBank mRNA BC047949.1 - 3 399
BC137087.1 - 3 273 9021084
BC137088.1 - 3 273 9021085
CN482844.1 - 6 600

View Full Product Details