Gene Transcripts

Gene Symbol
SPINT2
Entrez Gene ID
10653
Gene Name
serine peptidase inhibitor, Kunitz type 2
Gene Aliases
DIAR3, HAI-2, HAI2, Kop, PB
Chromosome Location
Chr. 19: 38264458 - 38292614 on Build GRCh38
UniGene ID
Hs.31439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166103.1 NP_001159575.1 4 715
NM_021102.3 NP_066925.1 5 886
GenBank mRNA AB006534.1 - 5 763
AF027205.1 - 5 815
AK127479.1 - 1 1776
AK291532.1 - 5 701
AK297154.1 - 4 390
AX772830.1 - 4 597
BC001668.2 - 5 782 2959462
BC007705.2 - 5 730 3952243
BC011951.1 - 5 686 3856970
BC011955.1 - 5 658 3857238
BC012868.2 - 5 773 3857277
BT020115.1 - 5 451
BT020116.1 - 5 451
BT020117.1 - 5 451
CR407604.1 - 5 451
U78095.1 - 5 751

View Full Product Details