Gene Transcripts

Gene Symbol
RIPPLY2
Entrez Gene ID
134701
Gene Name
ripply transcriptional repressor 2
Gene Aliases
C6orf159, SCDO6, dJ237I15.1
Chromosome Location
Chr. 6: 83853266 - 83857515 on Build GRCh38
UniGene ID
Hs.149454
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009994.2 NP_001009994.1 3 365
NR_103525.1 2 271
GenBank mRNA AK124105.1 - 1 547
BC130460.1 - 3 330 40146491
BC132968.1 - 3 330 40146990
BM782014.1 - 2 224

View Full Product Details