Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177519.2 | NP_001170990.1 | 4 | 886 | |
NM_001289152.1 | NP_001276081.1 | 4 | 1750 | ||
NM_001289153.1 | NP_001276082.1 | 4 | 1730 | ||
NM_001289154.1 | NP_001276083.1 | 4 | 1558 | ||
GenBank mRNA | AK299135.1 | - | 4 | 858 |