Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012727.1 | NP_001012745.1 | 2 | 313 | |
NM_006412.3 | NP_006403.2 | 2 | 313 | ||
GenBank mRNA | AF000237.1 | - | 2 | 277 | |
AF011374.1 | - | 2 | 271 | ||
BC000026.2 | - | 2 | 256 | 3507150 | |
BC004529.2 | - | 2 | 247 | 3347049 | |
BC019292.1 | - | 2 | 265 | 2823580 | |
BF347518.1 | - | 2 | 313 | ||
CN407343.1 | - | 2 | 313 | ||
U56418.2 | - | 2 | 250 |