Gene Transcripts
- Gene Symbol
- GABRG2
- Entrez Gene ID
- 2566
- Gene Name
- gamma-aminobutyric acid type A receptor gamma2 subunit
- Gene Aliases
- CAE2, ECA2, GEFSP3
- Chromosome Location
- Chr. 5: 162067642 - 162155539 on Build GRCh38
- UniGene ID
- Hs.7195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000816.3 |
NP_000807.2
|
2, 3 |
611 |
|
NM_198903.2 |
NP_944493.2
|
2, 3 |
611 |
|
NM_198904.2 |
NP_944494.1
|
2, 3 |
611 |
|
GenBank mRNA |
AK299640.1 |
- |
3, 4 |
336 |
|
AK315200.1 |
- |
2, 3 |
263 |
|
BC059389.1 |
- |
2, 3 |
432 |
5288256 |
BC069348.1 |
- |
2, 3 |
347 |
7262321 |
BC074795.2 |
- |
2, 3 |
285 |
30915258 |
CD013896.1 |
- |
2, 3 |
389 |
|
CD013897.1 |
- |
2, 3 |
383 |
|
X15376.1 |
- |
2, 3 |
479 |
|
View Full Product Details