Gene Transcripts

Gene Symbol
SCARF2
Entrez Gene ID
91179
Gene Name
scavenger receptor class F member 2
Gene Aliases
NSR1, SREC-II, SREC2, SRECRP-1, VDEGS
Chromosome Location
Chr. 22: 20424584 - 20437859 on Build GRCh38
UniGene ID
Hs.474251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153334.6 NP_699165.3 11 3026
NM_182895.4 NP_878315.2 11 3011
GenBank mRNA AB024433.1 - 11 2977
AB052951.1 - 11 2917
AF088043.1 - 1 162
AF522196.1 - 11 3023
BC000584.2 - 2 1037
BC009326.2 - 8 2094

View Full Product Details