Gene Transcripts
- Gene Symbol
- TMEM67
- Entrez Gene ID
- 91147
- Gene Name
- transmembrane protein 67
- Gene Aliases
- JBTS6, MECKELIN, MKS3, NPHP11, TNEM67
- Chromosome Location
- Chr. 8: 93754844 - 93832653 on Build GRCh38
- UniGene ID
- Hs.116240
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142301.1 |
NP_001135773.1
|
19, 20 |
1871 |
|
NM_153704.5 |
NP_714915.3
|
18, 19 |
1940 |
|
NR_024522.1 |
|
18, 19 |
1940 |
|
XM_006716686.3 |
XP_006716749.1
|
17, 18 |
1778 |
|
XM_011517363.2 |
XP_011515665.1
|
9, 10 |
1030 |
|
GenBank mRNA |
AK026742.1 |
- |
13, 14 |
1246 |
|
AK092244.1 |
- |
19, 20 |
1828 |
|
AK094935.1 |
- |
18, 19 |
1864 |
|
AK297676.1 |
- |
3, 4 |
593 |
|
AK308735.1 |
- |
9, 10 |
1007 |
|
BC032835.2 |
- |
18, 19 |
1858 |
4825770 |
BC054338.1 |
- |
18, 19 |
1867 |
|
BX648768.1 |
- |
18, 19 |
1884 |
|
KU178827.1 |
- |
18, 19 |
1839 |
|
KU178828.1 |
- |
17, 18 |
1685 |
|
View Full Product Details