Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135031.1 | NP_001128503.1 | 3 | 324 | |
NM_004188.6 | NP_004179.3 | 7 | 998 | ||
XM_006717297.3 | XP_006717360.1 | 3 | 489 | ||
XM_011519068.2 | XP_011517370.1 | 5 | 889 | ||
XM_011519069.2 | XP_011517371.1 | 4 | 659 | ||
XM_011519070.2 | XP_011517372.1 | 5 | 889 | ||
XM_017015175.1 | XP_016870664.1 | 4 | 619 | ||
XM_017015176.1 | XP_016870665.1 | 5 | 889 | ||
GenBank mRNA | AF081946.1 | - | 2 | 173 | |
AY428733.1 | - | 3 | 324 | ||
BC043371.1 | - | 7 | 998 | 5170920 | |
BC048254.1 | - | 2 | 204 | ||
CR536546.1 | - | 2 | 173 |