Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005554.3 | NP_005545.1 | 9 | 2384 | |
GenBank mRNA | BC008807.2 | - | 9 | 2223 | |
BC014152.2 | - | 9 | 2227 | 3639270 | |
BC069269.1 | - | 9 | 2205 | 5428084 | |
BG059253.1 | - | 1 | 67 |