Gene Transcripts

Gene Symbol
G6PC3
Entrez Gene ID
92579
Gene Name
glucose-6-phosphatase catalytic subunit 3
Gene Aliases
SCN4, UGRP
Chromosome Location
Chr. 17: 44070700 - 44076344 on Build GRCh38
UniGene ID
Hs.294005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028581.1 2 481
XM_011525473.2 XP_011523775.1 2 51
XM_011525474.2 XP_011523776.1 2 520
XM_017025335.1 XP_016880824.1 1 42

View Full Product Details