Gene Transcripts

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
Chromosome Location
Chr. 17: 18108706 - 18180227 on Build GRCh38
UniGene ID
Hs.462390
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016239.3 NP_057323.3 17 5283
XM_011523918.2 XP_011522220.1 15 5637
XM_017024714.1 XP_016880203.1 15 5637
XM_017024715.1 XP_016880204.1 15 5646

View Full Product Details