Gene Transcripts

Gene Symbol
NDUFAF2
Entrez Gene ID
91942
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 2
Gene Aliases
B17.2L, MMTN, NDUFA12L, mimitin
Chromosome Location
Chr. 5: 60945129 - 61153037 on Build GRCh38
UniGene ID
Hs.591757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174889.4 NP_777549.1 2 284
GenBank mRNA AB183433.1 - 2 157
AF087990.1 - 2 253
AK026051.1 - 2 284
AK291296.1 - 2 247
BC001753.1 - 1 206 3354845
BC022869.1 - 2 185
BC033965.1 - 2 200 5288040
KU178834.1 - 2 157
KU178835.1 - 2 157
KU178836.1 - 2 157

View Full Product Details