Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171993.1 | NP_001165464.1 | 12, 13 | 1282 | |
NM_002150.2 | NP_002141.1 | 10, 11 | 806 | ||
GenBank mRNA | AK057510.1 | - | 12, 13 | 1282 | |
AK290826.1 | - | 10, 11 | 802 | ||
BC024287.1 | - | 10, 11 | 788 | 5087393 | |
X72389.1 | - | 10, 11 | 791 |