Gene Transcripts

Gene Symbol
ANO10
Entrez Gene ID
55129
Gene Name
anoctamin 10
Gene Aliases
SCAR10, TMEM16K
Chromosome Location
Chr. 3: 43366324 - 43691594 on Build GRCh38
UniGene ID
Hs.656657
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204831.1 NP_001191760.1 9 1577
NM_001204832.1 NP_001191761.1 8 1379
NM_001204833.1 NP_001191762.1 7 1244
NM_001204834.1 NP_001191763.1 8 1007
NM_018075.3 NP_060545.3 9 1577
XM_011533885.2 XP_011532187.2 9 1543
XM_011533887.2 XP_011532189.1 9 1543
XM_011533889.2 XP_011532191.1 9 1543
XM_011533890.2 XP_011532192.1 9 1543
XM_017006716.1 XP_016862205.1 9 1543
XM_017006717.1 XP_016862206.1 9 1495
XM_017006718.1 XP_016862207.1 9 1522
XM_017006719.1 XP_016862208.1 8 1297
XM_017006720.1 XP_016862209.1 9 1522
XM_017006721.1 XP_016862210.1 8 1306
XM_017006722.1 XP_016862211.1 8 1279
GenBank mRNA AK096302.1 - 9 1577
AK131223.1 - 8 1295
AK292368.1 - 9 1462
AK295969.1 - 7 1179
AK297949.1 - 9 1520
AL832508.1 - 9 1444
BC038855.1 - 8 905

View Full Product Details