Gene Transcripts
- Gene Symbol
- WDR38
- Entrez Gene ID
- 401551
- Gene Name
- WD repeat domain 38
- Gene Aliases
- Chromosome Location
- Chr. 9: 124853347 - 124857891 on Build GRCh38
- UniGene ID
- Hs.343383
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001045476.2 |
NP_001038941.1
|
7, 8 |
887 |
|
NM_001276374.1 |
NP_001263303.1
|
7, 8 |
887 |
|
NM_001276375.1 |
NP_001263304.1
|
7, 8 |
854 |
|
NM_001276376.1 |
NP_001263305.1
|
6, 7 |
834 |
|
XM_005251987.3 |
XP_005252044.1
|
6, 7 |
876 |
|
GenBank mRNA |
BC127949.1 |
- |
7, 8 |
851 |
|
BC150645.1 |
- |
7, 8 |
887 |
9057015 |
BC171828.1 |
- |
7, 8 |
887 |
9054482 |
BC171830.1 |
- |
7, 8 |
854 |
9054484 |
BC171837.1 |
- |
6, 7 |
834 |
|
View Full Product Details