Gene Transcripts

Gene Symbol
NBPF10
Entrez Gene ID
100132406
Gene Name
neuroblastoma breakpoint family member 10
Gene Aliases
AB6, AG1, NBPF9
Chromosome Location
Chr. 1: 146066318 - 146144804 on Build GRCh38
UniGene ID
Hs.714127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK000726.1 - 2 757
BC021111.1 - 2 184
BC063799.1 - 2 167
BX538005.1 - 7 1275

Gene Symbol
LOC105371303
Entrez Gene ID
105371303
Gene Name
neuroblastoma breakpoint family member 10-like
Gene Aliases
Chromosome Location
Chr. 1: 145395130 - 145425602 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003020.1 XP_016858509.1 6 1068

Gene Symbol
NBPF20
Entrez Gene ID
100288142
Gene Name
neuroblastoma breakpoint family member 20
Gene Aliases
Chromosome Location
Chr. 1: 145289900 - 145405778 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278267.1 NP_001265196.1 237

Gene Symbol
NBPF9
Entrez Gene ID
400818
Gene Name
neuroblastoma breakpoint family member 9
Gene Aliases
AE01
Chromosome Location
Chr. 1: 149054033 - 149090861 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037675.3 NP_001032764.2 2 180
NM_001277444.1 NP_001264373.1 2 180
XM_006711317.2 XP_006711380.1 3 408
XM_017001278.1 XP_016856767.1 3 408
GenBank mRNA AY894574.1 - 2 182

View Full Product Details