Gene Transcripts

Gene Symbol
TRIM67
Entrez Gene ID
440730
Gene Name
tripartite motif containing 67
Gene Aliases
TNL
Chromosome Location
Chr. 1: 231162928 - 231221568 on Build GRCh38
UniGene ID
Hs.655089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004342.3 NP_001004342.3 2, 3 1181
NM_001300889.1 NP_001287818.1 4, 5 995
XM_011544192.2 XP_011542494.1 2, 3 1181
XM_017001323.1 XP_016856812.1 2, 3 1181
GenBank mRNA AK126782.1 - 2, 3 1181
AY253917.1 - 4, 5 953

View Full Product Details