Gene Transcripts

Gene Symbol
OR2J3
Entrez Gene ID
442186
Gene Name
olfactory receptor family 2 subfamily J member 3
Gene Aliases
6M1-3, C3HEXS, HS6M1-3, OR6-16, OR6-6, OR6.3.6, ORL671
Chromosome Location
Chr. 6: 29111810 - 29112884 on Build GRCh38
UniGene ID
Hs.738947
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005216.3 NP_001005216.2 1 645
GenBank mRNA BC136973.1 - 1 645
BC136975.1 - 1 646 9020967

View Full Product Details