Gene Transcripts
- Gene Symbol
- CBWD6
- Entrez Gene ID
- 644019
- Gene Name
- COBW domain containing 6
- Gene Aliases
- Chromosome Location
- Chr. 9: 41131309 - 41198865 on Build GRCh38
- UniGene ID
- Hs.597638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001085457.1 |
NP_001078926.1
|
8, 9 |
764 |
|
XM_017015019.1 |
XP_016870508.1
|
7, 8 |
764 |
|
XM_017015020.1 |
XP_016870509.1
|
7, 8 |
764 |
|
XM_017015021.1 |
XP_016870510.1
|
7, 8 |
764 |
|
XM_017015023.1 |
XP_016870512.1
|
7, 8 |
764 |
|
XM_017015026.1 |
XP_016870515.1
|
7, 8 |
764 |
|
XM_017015027.1 |
XP_016870516.1
|
7, 8 |
764 |
|
XM_017015028.1 |
XP_016870517.1
|
8, 9 |
632 |
|
XM_017015029.1 |
XP_016870518.1
|
4, 5 |
583 |
|
XM_017015030.1 |
XP_016870519.1
|
7, 8 |
764 |
|
XM_017015031.1 |
XP_016870520.1
|
3, 4 |
275 |
|
XM_017015032.1 |
XP_016870521.1
|
7, 8 |
764 |
|
XM_017015034.1 |
XP_016870523.1
|
2, 3 |
138 |
|
XM_017015035.1 |
XP_016870524.1
|
8, 9 |
824 |
|
View Full Product Details