Gene Transcripts

Gene Symbol
TRIM64C
Entrez Gene ID
646754
Gene Name
tripartite motif containing 64C
Gene Aliases
Chromosome Location
Chr. 11: 49053714 - 49059112 on Build GRCh38
UniGene ID
Hs.740240
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206631.1 NP_001193560.1 4, 5 753

View Full Product Details