Gene Transcripts

Gene Symbol
MYL5
Entrez Gene ID
4636
Gene Name
myosin light chain 5
Gene Aliases
Chromosome Location
Chr. 4: 674199 - 682033 on Build GRCh38
UniGene ID
Hs.410970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002477.1 NP_002468.1 2, 3 226
XM_006713886.2 XP_006713949.1 2, 3 603
XM_017008245.1 XP_016863734.1 3, 4 1904
XM_017008246.1 XP_016863735.1 4, 5 1773
XM_017008247.1 XP_016863736.1 2, 3 1363
XM_017008248.1 XP_016863737.1 3, 4 386
GenBank mRNA AK123974.1 - 3, 4 2522
AK125529.1 - 3, 4 1206
BC040050.1 - 2, 3 346 6014371
L03785.1 - 2, 3 226

View Full Product Details