Gene Transcripts

Gene Symbol
MYL5
Entrez Gene ID
4636
Gene Name
myosin light chain 5
Gene Aliases
Chromosome Location
Chr. 4: 674199 - 682033 on Build GRCh38
UniGene ID
Hs.410970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002477.1 NP_002468.1 2 172
XM_006713886.2 XP_006713949.1 2 549
XM_017008245.1 XP_016863734.1 3 1850
XM_017008246.1 XP_016863735.1 4 1719
XM_017008247.1 XP_016863736.1 2 1309
XM_017008248.1 XP_016863737.1 3 332
GenBank mRNA AK123974.1 - 3 2468
AK125529.1 - 3 1152
BC040050.1 - 2 292 6014371
L03785.1 - 2 172

View Full Product Details