Gene Transcripts

Gene Symbol
CRLF2
Entrez Gene ID
64109
Gene Name
cytokine receptor-like factor 2
Gene Aliases
CRL2, CRLF2Y, TSLPR
Chromosome Location
Chr. X: 1190437 - 1212762 on Build GRCh38
UniGene ID
Hs.287729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022148.3 NP_071431.2 2, 3 271
NR_110830.1 2, 3 268
XM_011546181.2 XP_011544483.1 2, 3 306
XM_011546182.2 XP_011544484.1 2, 3 310
GenBank mRNA AB052639.1 - 2, 3 182
AF142570.1 - 2, 3 198
AF338733.1 - 2, 3 182
AK026800.1 - 2, 3 270
AK223303.1 - 2, 3 298
AK225583.1 - 2, 3 270

View Full Product Details