Gene Transcripts

Gene Symbol
GNG11
Entrez Gene ID
2791
Gene Name
G protein subunit gamma 11
Gene Aliases
GNGT11
Chromosome Location
Chr. 7: 93921704 - 93926514 on Build GRCh38
UniGene ID
Hs.83381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004126.3 NP_004117.1 2 593
GenBank mRNA AF493878.1 - 2 159
AK312037.1 - 2 243
BC009709.1 - 2 510 3879341
BT007141.1 - 2 159
U31384.1 - 2 266

View Full Product Details